Learn More
Masterflex™ 3-way Stopcock
Description
Regulate flow without any leaking with these stopcocks. Cole-Parmer™ Three-Way Stopcocks feature a male luer lock, PC body and an HDPE stem. Handle rotates 180 degrees.
Warning: This product is not approved or intended for, and should not be used for medical, clinical, surgical or other patient oriented applications
Specifications
Specifications
| Type | Stopcock |
| Material | Polycarbonate, HDPE |
| Plug Material | Polycarbonate and HDPE |
| Outlet Port | Male Luer Lock |
| Connectors | Male Luer Lock |