Learn More
3M™ Exhalation Valve Cover for 7000 Series Respirator
Product Code. 15799827
Change view
Click to view available options
:
Gross wt.: 0.86 lb.; 20/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
15799827 Gross wt.: 0.86 lb.; 20/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15799827 Supplier 3M™ Supplier No. 7000088356
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Replacement part for 7800S full facepieces.
  • Designed for 3M Full Facepiece Respirator 7800S (sold separately)
  • Valve cover maintains original service performance of facepiece
  • 3M Catalog# 7918
TRUSTED_SUSTAINABILITY
Specifications
Product Type Exhalation Valve Cover
For Use With (Application) Assembly, blasting, cleaning, demolition, electrical, facility maintenance, grinding, health and safety testing, machine operation, painting, remediation, sanding, welding
For Use With (Equipment) Full facepiece 7800s
Model 7918
Dimensions (L x W x H) 10.1 x 7.9 x 4.4 in. (25.6 x 20 x 11.1 cm)
Length (English) 10.1 in.
Length (Metric) 25.6 cm
Width (English) 7.9 in.
Width (Metric) 20 cm
Height (English) 4.4 in.
Height (Metric) 11.1 cm
Weight (English) 0.86 lb.
Weight (Metric) 0.39 kg