Learn More
3M™ Petroleum Sorbent Pads
Description
- For use in oil-only sorbent applications
- May be used on work surface in anticipation of a spill
- Ideal for use under docks, in shallow water, rocky shorelines, marshes and beaches
- 3M Catalog# HP156 , 3M Catalog# T151
Specifications
Specifications
| Color | White |
| Capacity (English) | 43.5 gal. |
| Capacity (Metric) | 164.66 L |
| Type | Petroleum Sorbent Pad |
| For Use With (Application) | Pad may be used on a work surface in anticipation of a spill or as a wipe |
| Model | T151 |
| Dimensions (L x W) | 19 x 17 in. (48.2 x 43.18 cm) |
| Length (English) | 19 in. |
| Length (Metric) | 48.2 cm |
| Width (English) | 17 in. |
| Show More |