missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BioTek™ 800TS Absorbance Reader
For measuring absorbance in microplate-based assays
Brand: BioTek™ 800TS
Additional Details : Weight : 8.50000kg
Description
- For ELISA, protein, endpoint assays, kinetics, cell-based assays
- 4.3 in. color touchscreen LCD
- Linear shaking
- Gen5™ for reader control (Gen5™ IVD and Gen5™ Secure available separately)
- Tungsten halogen lamp
Warranty
12 months
Specifications
| 800TS Absorbance Microplate Reader | |
| Tungsten halogen lamp | |
| Filters | |
| 405, 450, 490, 630 | |
| Linear | |
| 8kg | |
| 100-240VAC at 50 to 60Hz | |
| CE and TUV marked. IVD compliant. |
| Absorbance | |
| End point, kinetic and well area scanning | |
| 400 to 750nm | |
| 10nm | |
| Type B for computer control, USB Type A for thumb drive and printer | |
| 100-240V | |
| 6- to 96-well plates | |
| Measuring absorbance in microplate-based assays |