Learn More
Eppendorf™ 96-Well Protein LoBind Deep Well Plates, White
Use these plates in proteomics or other fields of protein research, where protein concentration tend to be very small and sample recovery is vital for assay results. Eppendorf™ Deepwell Protein LoBind Plates recover significantly more protein for downstream analyses and enzymes remain active.
304.00€ - 1788.00€
Specifications
| Certified | Yes |
|---|---|
| Dnase Free | Yes |
| Detection Method | Quantitative Protein Analysis |
| For Use With (Application) | For PCR techniques |
| Max. Heat Stability | 121°C |
| Product Code | Brand | Volume (Metric) Well | No. per Pack | Color | Price | Quantity & Availability | |||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Volume (Metric) Well | No. per Pack | Color | Price | Quantity & Availability | |||
|
10709263
|
Eppendorf™
0030504208 |
1000 μL | 20/Pk | White |
304.00€
Pack of 20 |
|
10215373
|
Eppendorf™
0030508203 |
White |
1090.00€
Pack of 80 |
19-01-2021 |
10367133
|
Eppendorf™
0030508106 |
120 | White |
1788.00€
Pack of 120 |
19-01-2021 |
10574073
|
Eppendorf™
0030504119 |
Yellow |
663.00€
Pack of 40 |
19-01-2021 |
11518643
|
Eppendorf™
0030504305 |
20/Pk | White |
359.00€
Pack of 20 |
13-04-2021 |
Description
- LoBind material guarantees maximum sample recovery for improved assay results
- Free of surface coating (e.g., silicone) to minimize the risk of sample interference
- Lot-certified PCR clean purity grade: of human DNA, DNAase, RNase and PCR inhibitors
- Available in tube, microplate, and deepwell plate formats for easy-up scaling
- Unique OptiTrack™ matrix: 30% faster sample identification and fewer pipetting errors
- RecoverMax™ well design: optimized well geometry for minimal remaining/dead volume and excellent mixing properties
- Raised well rims and a smooth surface guarantee reliable sealing in plates
Specifications
| Yes | |
| Quantitative Protein Analysis | |
| 121°C |
| Yes | |
| For PCR techniques | |
| Deep well plates, 96 Wells |