Learn More
Eppendorf™ Adapter
Product Code. p-7113373 Shop All Eppendorf Products
Change view
Click to view available options
For Use With (Equipment):
Adapter 13x75-100mm round bottom/PL/TB
Adapter 15mL conical tubes, PL/TB
Adapter 16x75-100mm round bottom/PL/TB
Adapter 17.5x100mm round bottom/PL/TB
Adapter 50mL conical tubes, PL/TB
for 500mL Nalgene
for 600mL TPP
Unit Size:
Pack of 2
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. For Use With (Equipment) unitSize
15797918 for 600mL TPP Pack of 2
15738379 for 500mL Nalgene Pack of 2
15758379 Adapter 13x75-100mm round bottom/PL/TB Pack of 2
15768379 Adapter 16x75-100mm round bottom/PL/TB Pack of 2
15778379 Adapter 17.5x100mm round bottom/PL/TB Pack of 2
15788379 Adapter 50mL conical tubes, PL/TB Pack of 2
15798379 Adapter 15mL conical tubes, PL/TB Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Product Code. 15797918 Supplier Eppendorf™ Supplier No. 5920701002
Only null left
Add to basket
Add to basket
Adaptors available for a variety of Eppendorf products to suit your needs.
Adapter 600mL w. conical tubes/RB (2x)
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) for 600mL TPP
No. per Pack Set of 2