Description
- Rotor F-35-6-30 holds 6 x 15/50mL conical or 12 × Vacutainer™ tubes
Specifications
Specifications
| Type | Small Rotor Bore Adapter |
| Tube Type | Round Bottom |
| Capacity | 1 x 2.6 to 7 mL Tube |
| For Use With (Equipment) | Rotor F-35-6-30 |
| Bore Size (Metric) | Small rotor bore |
| No. per Pack | 2/Pk. |