Description
- Rotor F-35-6-30 holds 6 x 15/50mL conical or 12 × Vacutainer™ tubes
Specifications
Specifications
| Type | Large Rotor Bore Adapter |
| Tube Type | Round Bottom |
| Capacity | 6 x 50 mL |
| Max. Tube Diameter | 29 mm |
| For Use With (Equipment) | Rotor F-35-6-30 |
| Bore Size (Metric) | Large rotor bore |
| Description | 29 dia. x 95 to 125mm |
| No. per Pack | 2/Pk. |