Learn More
Eppendorf™ Adapter for S-4x400 Bucket
Product Code. p-7159250 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
1 x 175 to 225 mL Conical Tubes
1 x 175 to 250 mL Flat/Conical Bottle
1 x 400 mL Bottle
11 x 7.5 to 12 mL Round Bottom Tubes
15 x 4 to 8 mL Round Bottom Tubes
17 x 12 x 75 mm Round Bottom Tubes
26 x 1.5/2.0 mL Microtubes
4 x 50 mL Conical Tubes
50 mL Self-standing Tubes, 50 mL Centriprep, 30 mL Oak Ridge
7 x 5.0 mL Microtubes, 15 mL Conical Tubes
8 x 9 mL Round Bottom Tubes
Unit Size:
Pack of 2
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
15823232 1 x 175 to 225 mL Conical Tubes Pack of 2
15853222 1 x 175 to 250 mL Flat/Conical Bottle Pack of 2
15863222 1 x 400 mL Bottle Pack of 2
15813232 50 mL Self-standing Tubes, 50 mL Centriprep, 30 mL Oak Ridge Pack of 2
15813222 4 x 50 mL Conical Tubes Pack of 2
15823222 7 x 5.0 mL Microtubes, 15 mL Conical Tubes Pack of 2
15883222 8 x 9 mL Round Bottom Tubes Pack of 2
15843222 11 x 7.5 to 12 mL Round Bottom Tubes Pack of 2
15833222 15 x 4 to 8 mL Round Bottom Tubes Pack of 2
15803232 17 x 12 x 75 mm Round Bottom Tubes Pack of 2
15873222 26 x 1.5/2.0 mL Microtubes Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15823232 Supplier Eppendorf™ Supplier No. 5910714009
Only null left
Add to basket
Add to basket
Facilitates the use of specific sizes, shapes, and numbers of vessels in rotor buckets for S-4x400 rotor.
  • Suitable for S-4x400 Rotor buckets
  • Accommodates selected array of tubes or bottles
TRUSTED_SUSTAINABILITY
Specifications
Tube Type Conical
Capacity 1 x 175 to 225 mL Conical Tubes
Max. Tube Diameter 62 mm
Max. Tube Length 143 mm
For Use With (Equipment) S-4x400 Bucket
Vessels per Adapter 1
No. of Places 1
No. per Pack 2/Pk.