Description
- Suitable for S-4xUniversal rotor buckets
- Accommodates selected array of tubes, bottles, or plates
Specifications
Specifications
| Tube Type | Round Bottom |
| Capacity | 30 x 4 to 8 mL Round Bottom Tubes |
| Max. Tube Diameter | 13 mm |
| Max. Tube Length | 119 mm |
| For Use With (Equipment) | S-4xUniversal Rotor |
| No. of Places | 30 |
| No. per Pack | 2 |