Learn More
Eppendorf™ Adapters For Oak Ridge Tubes
Product Code. p-8003421 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
10 mL
16 mL
30 mL
35 mL
Unit Size:
Pack of 2
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
12376053 10 mL Pack of 2
11987065 16 mL Pack of 2
10098179 30 mL Pack of 2
10784872 35 mL Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 12376053 Supplier Eppendorf™ Supplier No. 5820719001
Only null left
Add to basket
Add to basket
Add flexibility to your liquid handling applications with these adapters
  • Adapters
  • For Eppendorf™ Oak Ridge Tubes
TRUSTED_SUSTAINABILITY
Specifications
Type Adapter
Capacity 10 mL
For Use With (Equipment) 10mL Oak Ridge Tubes
No. per Pack 2 Pack