Learn More
Eppendorf™ Adapters for Rotors
Product Code. p-7113374 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
2.6 to 8 mL
250 mL Flat, 175 to 225 mL Conical
250 mL Flat, 175 to 225 mL Conical, Nunc 250 mL Conical
5 mL FACS
5.5 to 12 mL
500 mL Corning Conical Bottle
500 mL Nalgene Flat Bottle
6 x MTP, DWP, PCR Plates
750 mL Bottle
9 mL
Tube Type:
Conical
Flat Bottom
Flat/Conical Bottom
Round Bottom
For Use With (Equipment):
Rotor S-4x1000, High-capacity Buckets
Rotor S-4xUniversal-Large
S-4xUniversal-Large, Universal Buckets
Unit Size:
Pack of 2
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity Tube Type For Use With (Equipment) unitSize
15758389 2.6 to 8 mL Round Bottom Rotor S-4x1000, High-capacity Buckets Pack of 2
15728399 2.6 to 8 mL Round Bottom Rotor S-4xUniversal-Large Pack of 2
15715882 5 mL FACS Round Bottom Rotor S-4xUniversal-Large Pack of 2
15718399 5.5 to 12 mL Round Bottom Rotor S-4xUniversal-Large Pack of 2
15778389 9 mL Round Bottom Rotor S-4x1000, High-capacity Buckets Pack of 2
15738399 250 mL Flat, 175 to 225 mL Conical Flat/Conical Bottom Rotor S-4xUniversal-Large Pack of 2
15748389 250 mL Flat, 175 to 225 mL Conical, Nunc 250 mL Conical Flat/Conical Bottom Rotor S-4x1000, High-capacity Buckets Pack of 2
15758399 500 mL Corning Conical Bottle Conical Rotor S-4xUniversal-Large Pack of 2
15768399 500 mL Nalgene Flat Bottle Flat Bottom Rotor S-4xUniversal-Large Pack of 2
15748399 750 mL Bottle Flat Bottom Rotor S-4xUniversal-Large Pack of 2
15638918 6 x MTP, DWP, PCR Plates Flat Bottom S-4xUniversal-Large, Universal Buckets Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15758389 Supplier Eppendorf™ Supplier No. 5920718002
Only null left
Add to basket
Add to basket
Adapters for Rotors are for use with a variety of Eppendorf™, Corning™ and Nalgene™ tubes and plates in different shapes and sizes.
Available for use with:
  • 2.6 to 8 mL, set of 2 pcs.
  • 250 mL flat, 175 to 225 mL conical, and Nunc™ 250 mL conical, set of 2 pcs.
    • Please note that these conical bottles need to be used together with a conical form insert provided by the manufacturer.
  • 49 x 2.6 to 8 mL, set of 2 pcs.
  • 5.5 to 12 mL, set of 2 pcs.
  • 500 mL Corning™, set of 2 pcs.
  • 500 mL Nalgene™, set of 2 pcs.
  • 50 mL, set of 2 pcs.
  • 5 mL FACS, set of 2 pcs.
  • 750 mL, set of 2 pcs.
  • 9 mL, set of 2 pcs.
  • Eppendorf Tubes™ 5.0 mL and 15 mL, set of 2 pcs.
  • MTP, DWP, PCR plates, set of 2 pcs.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Tube Type Round Bottom
Capacity 2.6 to 8 mL
For Use With (Equipment) Rotor S-4x1000, High-capacity Buckets
No. of Places 49