Learn More
Thermo Scientific™ Adapters for TX-150 Swinging Bucket Rotors
Assorted tube adapters
259.66€ - 288.00€
Specifications
For Use With (Equipment) 50mL Conical Buckets for TX-150 rotor
View More Specs
Products 9
Product Code Brand Tube Type Price Quantity & Availability  
Product Code Brand Tube Type Price Quantity & Availability  
15195243 View Documents Thermo Scientific™
75005735
100mL Round Bottom Open-Top Tube 288.00€
Pack of 4 View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15105253 View Documents Thermo Scientific™
75005739
5/7mL Blood Collection Tube 288.00€
Pack of 4 View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15145263 View Documents Thermo Scientific™
75005744
25mL Universal Flat/Skirted Tube 288.00€
Pack of 4 View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15135243 View Documents Thermo Scientific™
75005736
50mL Conical or Skirted Tube 288.00€
Pack of 4 View alternative products
Save Up to 
»
In Stock 5
Add to basket
15551510 View Documents Thermo Scientific™
75003504
14mL Urine Tube 288.00€
Pack of 4 View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15165243 View Documents Thermo Scientific™
75005740
3/5mL Blood Collection Tube 288.00€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15183263 View Documents Thermo Scientific™
75005737
11mL IVF Tube 288.00€
Pack of 4 View alternative products
Save Up to 
»
In Stock 1
Add to basket
15135263 View Documents Thermo Scientific™
75005743
1.5/2mL Microtube 288.00€
Pack of 4 View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15184233 View Documents Thermo Scientific™
75005736
50mL Conical or Skirted Tube This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
N/A View alternative products
Save Up to 
»
Specifications
Specifications
50mL Conical Buckets for TX-150 rotor