missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Sulfate, ACS, ≥99%, Ultrapure, Affymetrix/USB™
Used in purification of enzymes and other proteins by precipitation
51.94€ - 165.00€
Chemical Identifiers
| CAS | 7783-20-2 |
|---|---|
| Molecular Formula | H8N2O4S |
| Molecular Weight (g/mol) | 132.134 |
| MDL Number | MFCD00003391 |
| InChI Key | BFNBIHQBYMNNAN-UHFFFAOYSA-N |
| Synonym | ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate |
| PubChem CID | 6097028 |
| ChEBI | CHEBI:62946 |
| IUPAC Name | diazanium;sulfate |
| SMILES | [NH4+].[NH4+].[O-]S(=O)(=O)[O-] |
| Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
|
15815258
|
Thermo Scientific
J11254.A1 |
1kg | Plastic bottle |
51.94€
1kg |
|
||||
|
15825258
|
Thermo Scientific
J11254.A7 |
5kg | Plastic bottle |
165.00€
5kg |
|
||||
Chemical Identifiers
| 7783-20-2 | |
| 132.134 | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| 6097028 | |
| diazanium;sulfate |
| H8N2O4S | |
| MFCD00003391 | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| CHEBI:62946 | |
| [NH4+].[NH4+].[O-]S(=O)(=O)[O-] |
Specifications
| Ammonium Sulfate | |
| H8N2O4S | |
| MFCD00003391 | |
| BFNBIHQBYMNNAN-UHFFFAOYSA-N | |
| diazanium;sulfate | |
| 6097028 | |
| 132.14 | |
| ≥99% | |
| 1.769 | |
| 1.769 | |
| 1kg |
| 7783-20-2 | |
| (NH4)2SO4 | |
| ammonium sulfate, diammonium sulfate, ammonium sulphate, mascagnite, sulfuric acid diammonium salt, ammonium sulfate 2:1, ammoniumsulfate, actamaster, sulfuric acid, diammonium salt, diammonium sulphate | |
| [NH4+].[NH4+].[O-]S(=O)(=O)[O-] | |
| 132.134 | |
| CHEBI:62946 | |
| Powder | |
| ACS | |
| Plastic bottle | |
| ~280°C (decomposition) |