Learn More
Techne™ Dri-Block™ Aluminum Heating Block Insert
Precision-engineered aluminum heating blocks offer excellent temperature accuracy
Brand: Techne™ F4481
Additional Details : Weight : 1.00000kg
Description
- Versatile range of interchangeable heating blocks work with many size sample tubes and plates
- Heaters sold separately
Specifications
| Flat bottom wells and perimeter ridge | |
| Anodized Aluminum | |
| Dri-Block™ Insert |