missing translation for 'onlineSavingsMsg'
Learn More

HLA-DR Mouse anti-Human, PE-Cy7, Clone: L243 (G46-6), BD

Mouse Monoclonal Antibody

Brand:  BD Biosciences 560651

Additional Details : Weight : 0.01000kg

Product Code. 15810379

  • 309.00€ / Each


Estimated Shipment
15-06-2020
Add to basket

Description

Description

Antin++DR recognizes a human class II major histocompatibility complex (MHC) antigen. The antigen is a transmembrane glycoprotein composed of α- and β-subunits that have molecular weights of 36 and 27kDa, respectively. Antin++DR reacts with a nonpolymorphic HLA-DR epitope and does not cross-react with HLA-DQ or HLA-DP molecules.

Flow Cytometry

Specifications

Specifications

HLA-DR
L243 (G46-6)
Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Murine
50 Tests
Immunology
Primary
Human
Flow Cytometry
PE-Cyanine7
Affinity Purified
IgG2a, κ
RUO
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Monoclonal
Videos
Documents