Learn More
Azlon™ Spouted HDPE Bucket
Description
- Handle grip for easy lifting and carrying
- Choice of white or black
- Moulded graduation
Specifications
Specifications
| Product Type | Spouted Bucket |
| Material | HDPE |
| Autoclavable | Not Autoclavable |
| Color | Black |
| Diameter (Metric) Outer | 330 mm |
| Height (Metric) | 315 mm |
| Quantity | 1 Ea. |