missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ LB Broth, Lennox

Product Code. p-4869115
Change view
Click to view available options
Quantity:
500 g
2 kg
Packaging Type:
Bottle
Pail
Unit Size:
2kg
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Type unitSize
11788842 2 kg Pail 2kg
11798842 500 g Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11788842 Supplier BD Supplier No. 240210

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Used to propagate and maintain recombinant strains of Escherichia coli in molecular biology procedures. BD™ Difco™ LB Broth, Lennox is a nutritionally rich media developed by Lennox for growth of pure E. coli cultures.

The Difco™ LB Broth, Lennox media provide all of the nutritional requirements of organisms that have been derived from E. coli K12, which are deficient in B vitamin production. E. coli grows more rapidly in Lennox LB Broth than in conventional media because these media provide the cells with amino acids, nucleotide precursors, vitamins and other metabolites that the microorganism would otherwise have to synthesize.

LB Broth, Lennox contains ten times the sodium chloride level of Luria Broth Base, Miller and one half of that found in LB Broth, Miller. This allows the researcher to select the optimal salt concentration for a specific strain. If desired, the medium may be aseptically supplemented with glucose to prepare the complete medium described by Lennox.

  • Formulation: Tryptone 10.0g, Yeast Extract 5.0g, Sodium Chloride 5.0 g
  • Good cultural response to: Escherichia coli 33694 (HB101), Escherichia coli 39403 (JM103), Escherichia coli 53868 (DH5)
  • To prepare, mix 20g in 1L distilled water and boil for 1 minute
  • pH (3.5% solution, 25°C): 7.0 ± 0.2
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance CE
Color Light beige
Content And Storage Refrigerate at <30°C
Form Dehydrated Powder
Media Type LB Broth, Lennox
Product Type Microbiology Media
Formulation Tryptone 10g/L, Yeast extract 5g/L, Sodium Chloride 5g/L
For Use With (Application) For maintaining and cultivating recombinant strains of Escherichia coli
Packaging Type Pail
Quantity 2 kg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.