Learn More
Bel-Art™ Slide Boxes
Product Code. 15515614
Change view
Click to view available options
Slide Capacity:
100
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Slide Capacity unitSize
15515614 100 Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15515614 Supplier Bel-Art™ Supplier No. H445770000
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Will not warp, splinter or crack, and are unaffected by moisture. Slide boxes are compact, durable slide boxes to provide the maximum protection for 25 x 75 mm (1 x 3 in.) microscope slides.
  • Heavy-walled boxes are rugged and will not bend, split or chip
  • Boxes are unaffected by humidity and moisture and are thoroughly insect-proof
  • Easily identify slides with numbered index included on base and cover
Specifications
Type Slide Box
Slide Capacity 100
Device Holds 100
For Use With (Equipment) Microscope Slide
Includes Numbered index on base
Product Title
Selezionare un problema
Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.