Learn More
Bel Art™ cuvette Rack
Description
- These two-tier racks support cuvettes with a minimum of physical contact, greatly reducing the chance of damage
- Only the corners of the cuvettes touch the racks
- Accommodate 12 cuvettes of 10 mm light path
- Will not scratch quartz cuvettes
- Choose autoclavable or nonautoclavable
Specifications
Specifications
| Autoclavable | Autoclavable |
| Color | White |
| Holds | Light Path Cuvettes (10 mm), Light Path Cuvettes (12 mm) |
| Material | Polypropylene |
| Positions | 12 |
| Shape | Square |
| Stackable | Yes |
| For Use With (Equipment) | Cuvettes |
| Dimensions (L x W x H) | 4.5 x 4.375 x 4.375 in. (11.4 x 11.1 x 11.1 cm) |
| Length (Metric) | 11.4 cm |
| Show More |