Learn More
Thermo Scientific™ BIOShield™ 1000A Rotor Adapters
Product Code. p-4527819 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity:
15 mL Conical Tube
50 mL Conical Tube
Unit Size:
Pack of 4
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
11874501 15 mL Conical Tube Pack of 4
11864501 50 mL Conical Tube Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11874501 Supplier Thermo Scientific™ Supplier No. 75003642
Increase rotor versatility with Thermo Scientific™ BIOShield™ 1000A Rotor Adapters, which provide support for 15/50mL conical tubes.
Ordering Information:
A variety of other adapters (sold in sets of four unless otherwise noted) are available for use with both the BIOShield 1000A Rotor (75-003-603) and the TX-750 Swinging-Bucket Rotor with Rectangular Buckets (75-003-607/75-003-614). Contact your sales representative for more information.
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Tube Adapter
Capacity 15 mL Conical Tube