Learn More
Rogo Sampaic™ Borosilicate Glass Test Tubes
Product Code. p-8009969
Change view
Click to view available options
Diameter (Metric):
12 mm
16 mm
20 mm
Length (Metric):
120 mm
160 mm
200 mm
Unit Size:
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Diameter (Metric) Length (Metric) unitSize
11723923 16 mm 160 mm Pack of 100
11707045 12 mm 120 mm Pack of 100
11727045 20 mm 200 mm Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11723923 Supplier Rogo Sampaic™ Supplier No. BGB044
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Ideal for routine laboratory analysis and procedures. Rogo Sampaic™ Borosilicate Glass Test Tubes are manufactured from thick walled clear borosilicate glass.
  • Corrosion-free and resistant to thermal shock
  • Autoclavable
  • Mouth slightly annealed and rounded
Specifications
Capacity (Metric) 20 mL
Length (Metric) 160 mm
Autoclavable Autoclavable
Diameter (Metric) 16 mm