Learn More
Thermo Scientific™ Buckets and Lids for Thermo Scientific Sorvall™ Legend™ Mach 1.6™ 4-Place Swinging Bucket Rotor 75-002-000
Product Code. 10317552 Shop All Thermo Scientific Products
Change view
Click to view available options
Type:
Bucket
Cap
Capacity:
150 mL
400 mL
Not Applicable
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Capacity unitSize
10317552 Bucket 400 mL Each
11886510 Cap Not Applicable Each
10246872 Bucket 150 mL Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10317552 Supplier Thermo Scientific™ Supplier No. 75002002
Only null left
Add to basket
Add to basket
For use with Sorvall Legend Mach 1.6/1.6R Centrifuges when equipped with 4-Place Swinging Bucket Rotor 75-002-000.
Choose rectangular or round buckets.
  • Rectangular Bucket (75-002-001) capacity: 150mL
  • Round Bucket (75-002-002) capacity: 400mL
  • Aerosol-tight lids are suitable for biocontainment use
Order Info
Buckets and lids are sold separately. Buckets are sold individually; order correct quantity for your rotor. Lids are sold in pairs.
TRUSTED_SUSTAINABILITY
Specifications
For Use With (Equipment) TTH-400 swing bucket rotor
Capacity 400 mL
Type Bucket
Tube Size 400 mL
Description Round Bucket