missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Buffers for NucleoSpin miRNA Plasma Kits

Supplemental buffer solutions (e.g., lysis buffer, protein precipitation buffer) for use with NucleoSpin™ miRNA Plasma Kits for the rapid isolation of small RNA from from plasma and serum samples.

Brand:  Macherey-Nagel™ 740367.25

Additional Details : Weight : 0.02500kg

 View more versions of this product

Product Code. 15869077

  • 35.26€ / 25mL


Estimated Shipment
19-01-2021
Add to basket

Description

Description

Isolation of small RNA and DNA from plasma and serum

  • Efficient and selective isolation of small RNA from plasma or serum samples (patent pending)
  • Superior recovery of small RNA ≤ 18 nt
  • Optional on-column DNA digest for increased sensitivity in downstream applications
  • Simple and fast procedure – without phenol / chloroform
Specifications

Specifications

NucleoSpin™ miRNA Plasma Kit
Protein Precipitation Buffer MPP
0.1
Liquid
4.0 - 4.6
Colorless
Isolating small RNA from plasma and serum samples
7646-85-7
0.25
Acetic
Water
25 mL
Videos
SDS
Documents