Learn More
Carl Zeiss™ C-mount Camera Adapter
Adapts specific Zeiss microscopes for use with still or video camera
Brand: Carl Zeiss™ 000000-1069-414
Additional Details : Weight : 0.80000kg
Description
Adapts microscope for use with C-mount still or video cameras; 0.63X f/60mm interface
Accessory for:
- Stemi 2000 C /CS
- Axiostar
- Axioskop
- AxioImager D1
Specifications
| Camera adapter | |
| C mount camera | |
| Stemi™ 2000C/CS, Axiostar™, Axioskop™, AxioImager D1 |