Learn More
Zeiss™ Light Guide Holder for Stemi™ Mounts
Designed for use with Zeiss Microscopes
Brand: Zeiss™ 417085-9002-000
Additional Details : Weight : 0.50000kg
Description
Compatible Microscopes:
- Stemi 2000
- Stemi 2000 C
- Stemi 2000 CS
- Stemi 305
- Stemi 305 Demo
- Stemi 305 trino
- STEMI SV 11
- Stemi SV 11 Apo
- STEMI SV 6
- SteREO Discovery.V12
- SteREO Discovery.V8
Specifications
| Light guide holder for Stemi mounts | |
| Stemi™ mounts | |
| Light guide holder | |
| Light guide holder |