missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
- The kit allows the selection of CD3+ T cells directly from whole blood.
- CD3 Fab-Strep, human
- Buffer CI (10x)
- Biotin stock solution
- Fab-TACS™ Gravity Adapter
- Fab-TACS™ Flow Restrictor
- The Fab-TACS™ Gravity column is filled with a Strep-Tactin™ coated matrix made of cell-grade agarose.
- Strep-tagged Fab fragments (Fab-Streps) specifically bind to the matrix.
- Subsequently, whole blood or other blood preparations pass through the column.
- Target cells adhere to the matrix based on the exclusive binding of the Fab-Strep to the target cell.
- Non-target cells are efficiently washed away.
- In a final step, the addition of biotin causes the elution of the target cells and the Fab-Streps.
- After elution, the Fab-Streps self-dissociate from the cell surfaces.
- The label-free authentic target cells are now ready for further downstream applications.
Specifications
Specifications
| Product Type | Non-magnetic Cell Isolation Kit |
| Content And Storage | 2°C to 8°C |
| Donor Source | Human Whole Blood |
| For Use With (Application) | Isolation of label-free cells |
| Sufficient For | 60 mL whole blood |
| Quantity | 4 Columns, Human |
Safety and Handling
Product Identifier
- CD3 Fab-TACS Gravity Isolation Kit,Human single ma rker,4 columns,capacity 15ml whole blood each
- Warning
- LONG-TERM AQUATIC HAZARD Chronic 3
- H412-Harmful to aquatic life with long lasting effects.
- P273-Avoid release to the environment.
- MIXTURE LIST-Contain: sodium azide
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?