Specifications
| Product Type | Non-magnetic Cell Isolation Kit |
| Content And Storage | 2°C to 8°C |
| Donor Source | Mouse Spleen |
| For Use With (Application) | Isolation of label-free cells |
| Sufficient For | 6x107 target cells |
| Quantity | 1 Kit |