missing translation for 'onlineSavingsMsg'
Learn More

IBA Lifesciences CD3 Isolation Kit for FABian™, Mouse

Product Code. p-7160108 Shop All IBA Lifesciences Products
Change view
Click to view available options
Quantity:
1 Kit
10 Kits
Unit Size:
1 set
10 sets
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16285350 1 Kit 1 set
16295350 10 Kits 10 sets
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16285350 Supplier IBA Lifesciences Supplier No. 66504001

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

For automated positive cell separation. The Isolation Kit for FABian™ provides label-free target cells without the use of high affinity antibodies and magnetic beads

  • The kit is used for the selection of CD3+ T cells from mouse spleen in conjunction with the FABian&trade system.
  • It includes CD3 Fab-Strep (mouse), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian&trade run to isolate up to 6x10^7 target cells.
  • Specifications

    Product Type Non-magnetic Cell Isolation Kit
    Content And Storage 2°C to 8°C
    Donor Source Mouse Spleen
    For Use With (Application) Isolation of label-free cells
    Sufficient For 6x107 target cells
    Quantity 1 Kit
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.