Learn More
CD3 Mouse anti-Human, APC, Clone: HIT3A, BD
Mouse Monoclonal Antibody
185.00€ - 397.00€
Specifications
| Antigen | CD3 |
|---|---|
| Isotype | IgG2a, κ |
| Formulation | Aqueous buffered solution containing BSA and ≤0.09% sodium azide. |
| Conjugate | APC |
| Primary or Secondary | Primary |
Description
The HIT3a monoclonal antibody specifically binds to the human CD3 ε-chain, a 20 kDa subunit of the CD3/T cell antigen receptor complex found on 70-80% of normal human peripheral blood lymphocytes and 60-85% of thymocytes. Studies from the HLDA Workshop show that this antibody can be mitogenic for T lymphocytes. The CD3 complex plays a role in signal transduction during antigen recognition by the T cell receptor for antigen. HIT3a antibody does not stain intracellular CD3 unlike the other CD3-specific clone, UCHT1.
Specifications
| CD3 | |
| Aqueous buffered solution containing BSA and ≤0.09% sodium azide. | |
| Primary | |
| Murine | |
| Affinity Purified | |
| Monoclonal |
| IgG2a, κ | |
| APC | |
| Flow Cytometry | |
| RUO | |
| HIT3A | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
For Research Use Only.