missing translation for 'onlineSavingsMsg'
Learn More

IBA GmbH™ CD4 Isolation Kit for FABian™

IBA GmbH™ CD4 Isolation Kit for FABian™ is for reversible cell isolation delivering label-free target cells.

90.41€ - 718.00€

Specifications

Product Type Non-magnetic Cell Isolation Kit
Properties Isolation of label-free cells
Source Material Human whole blood
Storage Requirements 2°C to 8°C
Sufficient For 9mL whole blood
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15859774
View Documents
IBA GmbH™
6-6002-010
10 Kit, Human
718.00€
10 sets
View alternative products
Save Up to 
»
Estimated Shipment
18-12-2020
Add to basket
 
15849774
View Documents
IBA GmbH™
6-6002-001
1 Kit, Human
90.41€
1 set
View alternative products
Save Up to 
»
Estimated Shipment
18-12-2020
Add to basket
 
Description

Description

  • The kit is used for the selection of CD4+ T cells from whole blood or buffy coat in conjunction with the FABian™ system.
  • It includes CD4 Fab-Strep (human), the Cell Isolation Column, Cell Isolation Buffer, Biotin for Cell Elution and tubes and a luer lock adapter with the syringe.
  • Each kit is sufficient for one FABian™ run to isolate cells from 9 ml whole blood.
Specifications

Specifications

Non-magnetic Cell Isolation Kit
Human whole blood
9mL whole blood
Isolation of label-free cells
2°C to 8°C
SDS