Learn More
Sigma Laborzentrifugen™ Centrifuge Rotor Adapters
Securely hold glass and plastic tubes and bottles during the centrifuging process.
6.50€ - 283.00€
| Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Capacity | Price | Quantity & Availability | |||||
| 10187771 |
Sigma Laborzentrifugen™
13021 |
12 x .2mL |
6.50€ Pack of 2 |
|
11720829 |
Sigma Laborzentrifugen™
13059 |
1 x 10mL |
98.50€ Pack of 2 |
|
11740759 |
Sigma Laborzentrifugen™
16030 |
1 x 30mL |
55.30€ Pack of 2 |
|
13445195 |
Sigma Laborzentrifugen™
14177 |
1 x 175 to 250mL |
281.00€ Pack of 2 |
|
13435195 |
Sigma Laborzentrifugen™
14157 |
1 x 250mL |
283.00€ Pack of 2 |
|
11710719 |
Sigma Laborzentrifugen™
14150 |
2 x 50mL |
239.00€ Pack of 2 |
|
Description
- Hold a variety of tube sizes and capacities
- Adapters are compatible with Sigma rotors
Specifications
| 2/Pk. | |
| Tube Adapter |