Learn More
3M™ Chemical Spill Kit SK5
Product Code. 12634475
Change view
Click to view available options
Dimensions (L x W):
400 x 70 mm
Unit Size:
Pack of 5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Dimensions (L x W) unitSize
12634475 400 x 70 mm Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 12634475 Supplier 3M™ Supplier No. 7000088392
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
3M™ Hazardous Spill Response Kit is designed to clean up hazardous chemical spills quickly in an emergency situation.
  • Ideal for rapid response to chemical spills: fast and effective containment and clean-up
  • Cleaning up hazardous chemical spills, drips, and leaks
  • Suitable for use with most chemicals
  • Promotes safety on the site
  • Easy to dispose off
TRUSTED_SUSTAINABILITY
Specifications
Type Sorbent Spill Kit
Packaging Quantity 5/Cs.
Dimensions (L x W) 400 x 70 mm