Learn More
Welch™ CHEMSTAR™ Dry Vacuum Pump System
Product Code. 15516385 Shop All Welch Ilmvac Products
Change view
Click to view available options
Pump Type:
Vacuum System
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Pump Type unitSize
15516385 Vacuum System Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 15516385 Supplier Welch™ Supplier No. U2070C02
This item requires a price update prior to being ordered.
Request a Price for this item here.
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Integrated multistage pumping mechanism. Welch™ CHEMSTAR™ Dry Vacuum Pump System is software driven and fully chemical resistant.
  • Automated system
  • Pump corrosive gases and vapors
  • 50-millitorr with high flow
  • Oil-free, no cold trapping
  • Chemical resistant system: Compact and efficient, Very low maintenance
  • Eliminate pump oil changes and ritual of cold trapping
  • Electronically operated vapor load optimization and system self-cleaning
Applications:
  • Drying Manifolds (Schlenk lines)
  • Small freeze dryers
  • Small to medium vacuum ovens
  • Glove boxes
  • Concentrators/evaporators
  • Light industrial
TRUSTED_SUSTAINABILITY
Specifications
Amperage 3 A
Electrical Requirements 1 pH with Schuko cord set
Dimensions (L x W x H) 45.9 x 21.6 x 46.7 (18.1 x 8.5 x 18.4 in.)
Model 2070C-02
Weight (English) 50 lb.
Weight (Metric) 22.7 kg
Frequency 50/60 Hz
Pump Type Vacuum System
Maximum Pressure 0.07 mbar
Voltage 230 V