Learn More
Basic Line Conductivity Standard, 1.3μS/cm, Hamilton™
Product Code. 10661794
Change view
Click to view available options
Quantity:
250 mL
Unit Size:
300mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10661794 250 mL 300mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10661794 Supplier Hamilton Supplier No. 238973
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
Provides unprecedented stability. Hamilton™ Basic Line Conductivity Standard, 1.3μS/cm comes with a certificate with calibration document form DFM.
  • Remains stable for up to 1 year
  • Expiration date shown on every bottle
  • Bottles are permitted to stay open for a total of 60 minutes
Specifications
Packaging Glass bottle
Quantity 250 mL
Título del producto
Seleccione un problema
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.