Learn More
OHAUS™ Defender™ 5000 - D52 Low Profile Bench Scale, Models TD52P QR
Product Code. p-7162314 Shop All Ohaus Products
Change view
Click to view available options
Model:
D52P15RQDL5
D52P15RQDL5-M
D52P15RQDR5
D52P15RQDR5-M
D52P30RQDL5
D52P30RQDL5-M
D52P60RQDL5
D52P60RQDL5-M
D52P6RQDR5
D52P6RQDR5-M
Unit Size:
1 unit
10 product options available for selection
Product selection table with 10 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
15948565 D52P6RQDR5-M 1 unit
15958565 D52P15RQDR5-M 1 unit
15968475 D52P30RQDL5 1 unit
15968565 D52P15RQDL5-M 1 unit
15978455 D52P6RQDR5 1 unit
15978475 D52P60RQDL5 1 unit
15978565 D52P30RQDL5-M 1 unit
15988455 D52P15RQDR5 1 unit
15988565 D52P60RQDL5-M 1 unit
15998455 D52P15RQDL5 1 unit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 10 options available.
10 options
Product Code. 15948565 Supplier OHAUS™ Supplier No. 30461558
Add to basket
Add to basket
OHAUS™ Defender™ 5000 Series multifunctional bench scale is ideal for a multitude of applications. Built durably and equipped with advanced features such as maximum configurability print output, GMP/GLP data output, library/user management and multiple connectivity options.
  • User-friendly model with an easy-to-use keypad, large backlit display and plain text on-screen prompts which guide you through operation.
  • Durable construction with a stainless steel platform, sand gray powder-coated steel frame and aluminum load cell.
  • Defender meets NTEP/Measurement Canada or OIML/EC type approvals (depending on region).
  • Features common industrial modes and supports a 50000-item library with 1000 user profiles for easy data management.
  • Includes GMP/GLP with time/date for result traceability.
TRUSTED_SUSTAINABILITY
Specifications
Battery Life 20 Hours with Rechargeable Battery
Connection Type Easy access communication ports including RS232 and Micro SD slot (included), second RS232/RS485/USD device, Ethernet and WiFi/Bluetooth, Discrete I/O and Analog Output (sold separately as accessories); GLP/GMP data output with real-time clock
Construction ABS plastic (TD52P) indicator, 304-grade stainless steel platform with sand gray powder-coated steel frame, aluminum load cell, non-slip adjustable rubber feet
Capacity 3.00 kg, 6.00 kg
Display Type Backlit LCD
Readability 1.000000 g, 2.000000 g
Model D52P6RQDR5-M
Temperature (Metric) Operating -10°C to +40°C
Weighing Modes Gram, Kilogram, Metric Tonnes
Dimensions (L x W x H) 588 x 305 x 118 mm
Weight (Metric) 8.940 kg