Learn More
Techne™ Dri-Block™ Flat Bottom Plate Insert
Techne™ Dri-Block™ Flat Bottom Plate Insert for use with Techne™ Dri-Block™ Heaters
Brand:  Techne™ F4678
Additional Details : Weight : 4.00000kg
Product Code. 16009321
  • 460.00€ / Each
Estimated Shipment
23-02-2021
Add to basket
Description
Description
  • For use with Techne™ dri-block heaters
  • Manufactured from anodised aluminium
  • Blocks can be used in any combination
Specifications
Specifications
Flat-Bottom
(95 x 150 x 50 mm)
Anodized Aluminum
Techne™ Dri-Block Heaters