missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel Bioanalysis™ U-Bottom Elution Plate

Product Code. 11902432
Change view
Click to view available options
Quantity:
24/Pk.
Unit Size:
Pack of 24
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11902432 24/Pk. Pack of 24
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11902432 Supplier Macherey-Nagel Bioanalysis™ Supplier No. 740486.24
Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

96‐well microplates with 300 μL U‐bottom wells.

  • U-shaped bottom
  • Includes self-adhering foils

Specifications

Format Block
Content And Storage 18°C to 25°C
Quantity 24/Pk.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.