missing translation for 'onlineSavingsMsg'
Learn More

Eosin Y (Disodium Salt), Fisher BioReagents

Product Code. p-25710
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass Bottle
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10031503 25 g Glass Bottle 25g
10479814 100 g Glass Bottle 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10031503 Supplier Fisher BioReagents Supplier No. BP241925
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy
Eosin Y is used in microbiological differential media or as a biological stain.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17372-87-1
Molecular Formula C20H12Br4Na2O8
Molecular Weight (g/mol) 745.904
InChI Key MASXMTNVNASWNH-UHFFFAOYSA-L
Synonym Eosine Yellowish, Acid Red 87
PubChem CID 91886399
IUPAC Name disodium;2-(2,4,5,7-tetrabromo-3-oxido-6-oxoxanthen-9-yl)benzoate;trihydrate
SMILES C1=CC=C(C(=C1)C2=C3C=C(C(=O)C(=C3OC4=C(C(=C(C=C24)Br)[O-])Br)Br)Br)C(=O)[O-].O.O.O.[Na+].[Na+]

Specifications

Melting Point 300°C
Bio. Stain Comm. Certification Pass Test
Dye Content 88% min.
Assay Percent Range ≥88%
Merck Index 15, 3658
Formula Weight 691.85
Packaging Glass Bottle
Density 1.018g/cm³
Color Index No. 45380
Color Brown/Red
pH 7 to 10
Quantity 25 g
Physical Form Powder
Chemical Name or Material Eosin Y, Disodium Salt
Show More Show Less
compliance-icons
Product Identifier
  • Eosin Y
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.