Learn More
Eppendorf™ FA-45-6-30 Rotor Adapters
Product Code. 13246599 Shop All Eppendorf Products
Change view
Click to view available options
Capacity:
1 x 5 mL Conical Tube
1 x 5 mL Tube
13 x 100 mm Tubes
13 x 75 mm Tubes
16 x 100 mm Tubes
16 x 75 mm Tubes
Unit Size:
Pack of 2
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity unitSize
13246599 13 x 100 mm Tubes Pack of 2
13266599 16 x 100 mm Tubes Pack of 2
13256599 13 x 75 mm Tubes Pack of 2
13276599 16 x 75 mm Tubes Pack of 2
13286599 1 x 5 mL Tube Pack of 2
15158364 1 x 5 mL Conical Tube Pack of 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 13246599 Supplier Eppendorf™ Supplier No. 5820725001
Only null left
Add to basket
Add to basket
For volumes up to 50mL
  • FA-45-6-30 Rotor Adapters
  • For volumes up to 50mL
TRUSTED_SUSTAINABILITY
Specifications
Type Rotor Adapter
Capacity 13 x 100 mm Tubes
For Use With (Equipment) Rotor for 5804/5810 centrifuge
Vessels per Adapter 1 Tube