Includes
2 piece set
- For use with Centrifuge 5810/5810 R
Specifications
Specifications
| Type | Rotor Adapter |
| Capacity | 20 x 5.5/12 mL Tubes |
| For Use With (Equipment) | S-4-104 rotor for 5810/5810R centrifuge |
| No. per Pack | 2 Pack |