Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Filter aid, Celite Standard Super-cel

Click to view available options
Quantity:
500g
2500g
61790-53-2
O2Si
60.083
MFCD00132803
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O
61790-53-2
O2Si
60.083
MFCD00132803
VYPSYNLAJGMNEJ-UHFFFAOYSA-N
24261
CHEBI:30563
dioxosilane
O=[Si]=O
Applications
Filter aid, Celite Standard Super-cel is used to maximize biodiesel yields as a result of improved filtration, dewatering, dewaxing and winterization.

Solubility
Insoluble in water.

Notes
Incompatible with acids.
Quantity 500g
Odor Odorless
Merck Index 14,4973
Solubility Information Insoluble in water.
Chemical Name or Material Filter aid, Celite Standard Super-cel

GHS H Statement
H350-H373-H319-H335
May cause cancer.
May cause damage to organs through prolonged or repeated exposure.
Causes serious eye irritation.
May cause respiratory irritation.

P260-P264b-P271-P280i-P304+P340-P305+P351+P338-P312-P314-P501c

H319-H335-H373

EINECSNumber : 319-127-6

RTECSNumber : VV7311000

TSCA : Yes

Recommended Storage : Ambient temperatures