Learn More
Fisherbrand™ Free-Standing Microcentrifuge Tubes with Screw Caps
Autoclavable polypropylene
Specifications
| Autoclavable | Yes |
|---|---|
| Closure Material | Polypropylene |
| Closure Type | Screw |
| Color | Natural |
| Material | Polypropylene |
Description
Autoclavable polypropylene
- Withstand a maximum force of 20,000 × G
- Free-standing with cap
- Natural color
Order Info
500 per pack, 10 packs per case
Specifications
| Yes | |
| Screw | |
| Polypropylene |
| Polypropylene | |
| Natural | |
| Yes |