Learn More
Hartmann Larochette™ Foliodress™ Non-woven Surgical Face Masks
Product Code. p-8009430
Change view
Click to view available options
Color:
Blue
Green
Unit Size:
Pack of 50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Color unitSize
11730104 Blue Pack of 50
11720104 Green Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11730104 Supplier Hartmann Larochette™ Supplier No. 992537
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
High-quality surgical masks made of hyoallergenic, non-woven materials. Hartmann Larochette™ Foliodress™ Non-woven Surgical Face Masks are ideal for single use in surgical areas, isolation wards, and intensive care departments.
  • High-quality surgical face masks according to EN 14683:2014 type II or type II-R.
  • Hypoallergenic non-woven materials, free from natural rubber latex.
  • High filtration performance and comfortable breathing resistance
  • Long integrated nose bar.
  • Fragrance free.
  • Durable.
Specifications
Product Type Surgical Face Mask
Color Blue
Sterility Non-sterile
Certifications/Compliance EN 14683:2014 type II or type II-R
Construction Non-woven
Disposable Single-use
Filter Efficiency Bacterial
For Use With (Application) For single-use in all surgical areas, as well as on isolation wards and intensive care departments.