Learn More
Thermo Scientific™ Forma™ Steri-Cycle™ i160 CO2 Incubator, 165 L, Electropolished Stainless Steel
CO2 Sensor: IR Sensor; O2 Range (Oxygen Concentration): Optional 1 to 21% or 5 to 90%; Temperature Range: Ambient +3°C to 55°C
Brand:  Thermo Scientific™ 51030534
Additional Details : Weight : 85.00000kg
 View more versions of this product
Product Code. 15383012
  • 11292.00€ / Each
Estimated Shipment
22-04-2021
Add to basket
Description
Description
Direct Heat Chamber 
  • 165L electropolished stainless steel chamber with compact footprint, easily stackable 
  • Adjustable, perforated shelving 
  • Reversible exterior door for added flexibility
THRIVE  Active Airflow  
  • In-chamber fan gently distributes clean, humidified air throughout the chamber ensuring  homogeneous conditions 
  • Fast recovery of all parameters ten minutes or less for stable culturing conditions 
  • Auto-stop function disables fan operation during door openings to minimize air exchange
ISO 5 HEPA Filtration 
  • In-chamber HEPA continuously filters the entire chamber air volume every 60 seconds
  • ISO 5 clean room air quality is achieved in the culture chamber within 5 minutes
  • Continuous protection by minimizing ability for contaminants to settle on interior surfaces
Protected Humidity Reservoir 
  • Directly heated water reservoir provides stable high relative humidity levels, preventing culture desiccation 
  • Condensation free inner chamber, prevents a breeding ground for contaminant
  • 5X faster humidity recovery than traditional water pan designs 
In situ sensors and probes
  • Dual temperature probes  for over temperature protection with operational back up
  • Choice of thermal conductivity (TC) with rh compensation or single beam, dual wavelength infrared (IR) CO2 sensor
  • O2 control ranges 1 to 21% (hypoxic) and 5 to 90% (hyperoxic)
Steri-Run, high temperature defection cycle 
  • Overnight cycle simplifies cleaning and eliminates the need for separate autoclaving of parts
  • 180°C cycle assures uniform 6-log sterilization on all chamber surfaces
Intuitive, interactive iCAN™ Touchscreen Control
  • Bright, door-mounted display for easy accessibility and viewing
  • Selectable languages simplify operation: English, Spanish, German, French, Italian, Japanese and Mandarin
Specifications
Specifications
CE, CSA
Single 165L incubator, standard model
Electropolished Stainless Steel
34.6 x 25.1 x 35.4 in. (88 x 63.7 x 90 cm)
<93% at 37°C
230 V
CO2 Incubator
1 to 20%
USB, and optional 4-20mA
165 L
22.7 x 18.5 x 23.9 in. (57.6 x 47 x 60.7 cm)
Ambient +3°C to 55°C
Covered integrated water reservoir
IR Sensor
Optional 1 to 21% or 5 to 90%
Videos
SDS
Documents
Warranty