Learn More
Fisherbrand™ Gamma Radiation Shields
Product Code. p-8009033 Shop All Fisherbrand Products
Change view
Click to view available options
Includes:
15° Angle
45° Angle
Small Fixed 45° Angle
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Includes unitSize
12357187 15° Angle Each
12337187 45° Angle Each
12337177 Small Fixed 45° Angle Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 12357187
Only null left
Add to basket
Add to basket
Gamma radiation shields with fixed 15 degree angles and a curved base of various dimensions, are ideal for protection from Gamma radiation.
  • Radiation shielding Cleaver Scientific for protection from Gamma radiation, shield, fixed 15 degree angle, curved base, dimensions 530h x 350w x 150d mm
  • Provides clear, non-distorted vision, reducing light images and shadows, especially when the user is in a sitting position
  • Supplied in small, medium and large sizes and with curved base 15cm-deep for use with safety trays or flat 30x30cm base for under-the-bench protection
  • A range of angles offers increased maneuverability, while clear optical acrylic aids visualization
  • Available in two sizes, with either a flat base to provide bench protection, or a curved base which fits conveniently over safety trays
Specifications
Includes 15° Angle
Product Type Gamma Radiation Shield