Learn More
3M™ Gas and Vapour Filters
Product Code. p-8006577
Change view
Click to view available options
Model:
6038
6051
6054
6055
6057
6059
6075
6099
Unit Size:
10 pairs
2 pairs
32 pairs
4 pairs
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model unitSize
12352339 6059 4 pairs
15749007 6099 2 pairs
15729067 6057 4 pairs
15789007 6054 32 pairs
16802016 6055 4 pairs
15769007 6075 4 pairs
15799007 6051 4 pairs
15759027 6038 10 pairs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Product Code. 12352339 Supplier 3M™ Supplier No. 7000034747
Only null left
Add to basket
Edge
Add to basket
3M™ Gas and Vapour Filters; reusable masks with a bayonet connection and are designed to optimise your field of vision. Our filters offer lightweight and well-balanced breathing protection for hazardous environments.
Colour-coded labels according to the EN14387 classification system to show the type of protection they provide.These lightweight filters combine protection with comfort, and they’re compatible with our range of half and full-face masks for safe working in hazardous environments. They offer low breathing resistance and good balance when connected to your mask, and they’re designed to optimise your field of vision.They can be clicked into place quickly and easily, and they can be used with any 3M™ Reusable Masks that feature a bayonet-style connection. They can also be used in combination with 3M™ Particulate Filters and 3M™ 501 Particle Filter Retainers.
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance EN14387:2004 + A1:2008
Connection Type Bayonet
Description Ammonia and derivatives, Organic vapors bp >65°C
Product Type Combined Gas / Vapour Respirator Filter
Filter Efficiency ABEK1
For Use With (Application) Acid cleaning, Metal pickling, Metal etching and use, Chemical manufacture and handling
For Use With (Equipment) Half and Full Facemasks
Model 6059
Quantity 8/Pk.
Weight (Metric) 100 g