missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Biochrom™ GeneQuant™ 1300 Spectrophotometers
Can be used with a wide variety of cuvettes or capillaries for sample volumes from 3μL to 2mL
6741.00€ - 7183.00€
Specifications
| Detector Type | CCD array |
|---|---|
| Lamp | Xenon |
| Electrical Requirements | 100 to 240V 50/60Hz |
| Warranty | 3 Year |
| Wavelength Range | 190 to 1100nm |
| Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Product Type | Price | Quantity & Availability | |||||
|
10564815
|
Biochrom™
80-2120-00 |
GeneQuant 1300 Spectrophotometer |
6741.00€
Each |
21-01-2021 |
|||||
|
10034834
|
Biochrom™
80-2120-01 |
GeneQuant 1300 Spectrophotometer with printer |
7183.00€
Each |
29-01-2021 |
10574815
|
Biochrom™
80-2120-02 |
GeneQuant 1300 Spectrophotometer with Bluetooth |
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance |
|
Description
The GeneQuant 1300 spectrophotometer is a compact instrument pre- programmed with a range of methods for the quantitation of proteins, nucleic acids, and bacterial cell cultures. Absorbance and concentration measurements can be made at any wavelength, providing flexibility for future applications.
- Built-in applications for cell density, nucleic acids, proteins, and enzyme kinetics
- cDNA application software for measuring incorporation of Cy™3 and Cy™5, or other dyes into purified microarray hybridization probes and PCR products
- Wavelength range of 190 to 1100nm, with wavelength scan from 200 to 900nm performed in less than 5 sec. with zoom facility, peak identification, and on-peak confirmation
- Visual inspection of a nucleic acid scan can identify the presence of impurities especially useful with RNA samples
- Storage for 90 user-defined methods with (optional) password protection
- Large, easy-to-read graphical display
- Optics with no moving parts for high-energy throughput combined with a long-life Xenon lamp
- Optional built-in printer or output data via USB to a PC for printing or storage
- Wireless bluetooth connectivity (optional)
Warranty
Three-year lamp warranty
Specifications
| CCD array | |
| 100 to 240V 50/60Hz | |
| 190 to 1100nm | |
| 4.5kg |
| Xenon | |
| 3 Year | |
| ±0.5nm |