Learn More
OHAUS™ Guardian 7000 Hotplate-Stirrer
Durable hotplate stirrers with superior safety features and intelligent performance
703.00€ - 1105.00€
Specifications
Plug Type EU
Frequency 50/60 Hz
Voltage 230 V
View More Specs
Products 3
Product Code Brand Model Price Quantity & Availability  
Product Code Brand Model Price Quantity & Availability  
16680482
View Documents
OHAUS™
30500603
e-G71HS07C
784.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment
29-12-2020
Add to basket
16690482
View Documents
OHAUS™
30500613
e-G71HS10C
1105.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment
29-12-2020
Add to basket
16600492
View Documents
OHAUS™
30500623
e-G71HSRDM
703.00€
Each
View alternative products
Save Up to 
»
Estimated Shipment
29-12-2020
Add to basket
Description
Description
  • Protect your lab with patented SmartPresence™ & SmartLink™ technologies. These exclusive features automatically shut off the heater if no one is detected after a user set "time out" period has passed.
  • An industry-leading early detection system that uses two independent safety controls to continuously monitor the electronics and shut off heating before an overtemperature condition occurs.
  • Precise stirring from 60 to 1600 rpm is made possible by the design of a powerful motor, strong magnet and software controlled ramp rate, ensuring secure magnetic coupling for viscous applications.
Specifications
Specifications
EU
50/60 Hz
230 V
SDS