Learn More
Nabertherm High-Temperature Furnace, 1600 HFL Series
Lined with robust refractory insulation. Compared with the fiber-insulated models of the HT product line, these furnaces are recommended when high charge weights have to be sintered.
22994.74€ - 56912.91€
Products 4
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
15978280 View Documents Nabertherm
HF0326QNA
16 L 22994.74€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15998280 View Documents Nabertherm
HF0426QNA
40 L 37534.63€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15918290 View Documents Nabertherm
HF0626QNA
64 L 47026.29€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
15938290 View Documents Nabertherm
HF1626QNA
160 L 56912.91€
Each View alternative products
Save Up to 
»
please call +351214253350
Add to basket
Description
Description
  • Maximum Temperature: 1600 °C
  • Robust refractory brick insulation and special backing insulation
  • Furnace floor made of lightweight refractory bricks accommodates high charge weights
  • Defined application within the constraints of the operating instructions
  • NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive