Learn More
Nabertherm High-Temperature Furnace, 1600 HFL Series
Lined with robust refractory insulation. Compared with the fiber-insulated models of the HT product line, these furnaces are recommended when high charge weights have to be sintered.
22994.74€ - 56912.91€
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
| 15978280 |
Nabertherm
HF0326QNA |
16 L |
22994.74€ Each |
|
15998280 |
Nabertherm
HF0426QNA |
40 L |
37534.63€ Each |
|
15918290 |
Nabertherm
HF0626QNA |
64 L |
47026.29€ Each |
|
15938290 |
Nabertherm
HF1626QNA |
160 L |
56912.91€ Each |
|
Description
- Maximum Temperature: 1600 °C
- Robust refractory brick insulation and special backing insulation
- Furnace floor made of lightweight refractory bricks accommodates high charge weights
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive